siRNA Details
| Gene Symbol: Trim13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012210.1 | NP_001012210.1 | 3 | 302 | ||
| XM_006252137.2 | XP_006252199.1 | 3 | 389 | |||
| XM_008770767.1 | XP_008768989.1 | 3 | 370 | |||
| XM_008770768.1 | XP_008768990.1 | 2 | 286 | |||
| GenBank mRNA | BC088425.1 | - | 3 | 302 | 7123847 | |

Trim13
Genomic Map