siRNA Details
| Gene Symbol: Scara3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108870.1 | NP_001102340.1 | 4 | 522 | ||
| GenBank mRNA | FQ211593.1 | - | 4 | 539 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scara3| Gene Symbol: Scara3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108870.1 | NP_001102340.1 | 4 | 522 | ||
| GenBank mRNA | FQ211593.1 | - | 4 | 539 | ||