siRNA Details
| Gene Symbol: Tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108873.1 | NP_001102343.1 | 6 | 838 | ||
| GenBank mRNA | FJ515907.1 | - | 7 | 840 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnfrsf10b| Gene Symbol: Tnfrsf10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108873.1 | NP_001102343.1 | 6 | 838 | ||
| GenBank mRNA | FJ515907.1 | - | 7 | 840 | ||