siRNA Details
| Gene Symbol: Nufip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007758.1 | NP_001007759.1 | 6 | 789 | ||
| GenBank mRNA | BC082111.1 | - | 6 | 789 | 7135272 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nufip1| Gene Symbol: Nufip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007758.1 | NP_001007759.1 | 6 | 789 | ||
| GenBank mRNA | BC082111.1 | - | 6 | 789 | 7135272 | |