siRNA Details
| Gene Symbol: Nsa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134415.2 | NP_599242.2 | 4 | 597 | ||
| GenBank mRNA | BC098657.1 | - | 4 | 588 | 7378282 | |
| BP486932.1 | - | 1 | 92 | |||
| DQ354588.1 | - | 4 | 587 | |||
| FQ212851.1 | - | 4 | 609 | |||
| FQ214845.1 | - | 4 | 616 | |||
| FQ225571.1 | - | 4 | 610 | |||
| FQ226635.1 | - | 4 | 624 | |||
| FQ229542.1 | - | 4 | 615 | |||

Nsa2
Genomic Map