siRNA Details
| Gene Symbol: Dzip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252465.2 | XP_006252527.1 | 5, 6 | 984 | ||
| XM_006252466.1 | XP_006252528.1 | 5, 6 | 857 | |||
| XM_008770948.1 | XP_008769170.1 | 4, 5 | 1037 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dzip1| Gene Symbol: Dzip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252465.2 | XP_006252527.1 | 5, 6 | 984 | ||
| XM_006252466.1 | XP_006252528.1 | 5, 6 | 857 | |||
| XM_008770948.1 | XP_008769170.1 | 4, 5 | 1037 | |||