siRNA Details
| Gene Symbol: Phf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012211.1 | NP_001012211.1 | 7 | 683 | ||
| XM_006252632.2 | XP_006252694.1 | 5 | 501 | |||
| GenBank mRNA | BC079288.1 | - | 7 | 683 | 7134345 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf7| Gene Symbol: Phf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012211.1 | NP_001012211.1 | 7 | 683 | ||
| XM_006252632.2 | XP_006252694.1 | 5 | 501 | |||
| GenBank mRNA | BC079288.1 | - | 7 | 683 | 7134345 | |