siRNA Details
| Gene Symbol: Calr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012212.1 | NP_001012212.1 | 3 | 567 | ||
| GenBank mRNA | BC079049.1 | - | 3 | 567 | 7115321 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Calr3| Gene Symbol: Calr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012212.1 | NP_001012212.1 | 3 | 567 | ||
| GenBank mRNA | BC079049.1 | - | 3 | 567 | 7115321 | |