siRNA Details
| Gene Symbol: Ssbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014236.1 | NP_001014258.1 | 13 | 946 | ||
| XM_006252912.2 | XP_006252974.1 | 14 | 1013 | |||
| XM_008771139.1 | XP_008769361.1 | 14 | 1015 | |||
| XM_008771140.1 | XP_008769362.1 | 13 | 959 | |||
| XM_008771141.1 | XP_008769363.1 | 14 | 1013 | |||
| XM_008771142.1 | XP_008769364.1 | 13 | 959 | |||
| GenBank mRNA | BC079147.1 | - | 13 | 946 | 7128392 | |

Ssbp4
Genomic Map