siRNA Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108874.2 | NP_001102344.1 | 5 | 365 | ||
| XM_006252913.1 | XP_006252975.1 | 4 | 417 | |||
| XM_008771143.1 | XP_008769365.1 | 4 | 385 | |||
| GenBank mRNA | BC161989.1 | - | 5 | 365 | 9032206 | |

Tmem161a
Genomic Map