siRNA Details
| Gene Symbol: Tmem161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108874.2 | NP_001102344.1 | 9 | 918 | ||
| XM_006252913.1 | XP_006252975.1 | 8 | 970 | |||
| XM_006252914.1 | XP_006252976.1 | 7 | 673 | |||
| XM_008771143.1 | XP_008769365.1 | 8 | 938 | |||
| GenBank mRNA | BC161989.1 | - | 9 | 918 | 9032206 | |

Tmem161a
Genomic Map