siRNA Details
Gene Symbol: Tmem161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108874.2 | NP_001102344.1 | 11 | 1117 | ||
XM_006252913.1 | XP_006252975.1 | 10 | 1169 | |||
XM_006252914.1 | XP_006252976.1 | 9 | 872 | |||
XM_008771143.1 | XP_008769365.1 | 10 | 1137 | |||
GenBank mRNA | BC161989.1 | - | 11 | 1117 | 9032206 |
