siRNA Details
| Gene Symbol: Palld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751585.3 | XP_003751633.1 | 2 | 1110 | ||
| GenBank mRNA | BC088409.1 | - | 2 | 1064 | 7189104 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Palld| Gene Symbol: Palld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751585.3 | XP_003751633.1 | 2 | 1110 | ||
| GenBank mRNA | BC088409.1 | - | 2 | 1064 | 7189104 | |