siRNA Details
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108875.2 | NP_001102345.2 | 7 | 855 | ||
XM_008771302.1 | XP_008769524.1 | 7 | 689 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sgcz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108875.2 | NP_001102345.2 | 7 | 855 | ||
XM_008771302.1 | XP_008769524.1 | 7 | 689 |