siRNA Details
| Gene Symbol: Sgcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108875.2 | NP_001102345.2 | 8 | 987 | ||
| XM_008771302.1 | XP_008769524.1 | 8 | 821 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sgcz| Gene Symbol: Sgcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108875.2 | NP_001102345.2 | 8 | 987 | ||
| XM_008771302.1 | XP_008769524.1 | 8 | 821 | |||