siRNA Details
| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037327.1 | NP_001032404.1 | 34 | 5534 | ||
| GenBank mRNA | DQ124115.1 | - | 34 | 5534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csmd1| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037327.1 | NP_001032404.1 | 34 | 5534 | ||
| GenBank mRNA | DQ124115.1 | - | 34 | 5534 | ||