siRNA Details
| Gene Symbol: Wdr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751595.3 | XP_003751643.1 | 13, 14 | 2062 | ||
| XM_008771249.1 | XP_008769471.1 | 14, 15 | 2138 | |||
| XM_008771250.1 | XP_008769472.1 | 14, 15 | 2138 | |||

Wdr17
Genomic Map