siRNA Details
| Gene Symbol: Rnf170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253329.2 | XP_006253391.1 | 8 | 1041 | ||
| XM_006253330.2 | XP_006253392.1 | 8 | 976 | |||
| XM_006253331.2 | XP_006253393.1 | 8 | 1036 | |||
| XM_006253332.2 | XP_006253394.1 | 8 | 1045 | |||
| XM_344558.7 | XP_344559.5 | 8 | 981 | |||

Rnf170
Genomic Map