siRNA Details
Gene Symbol: B4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031661.1 | NP_001026831.1 | 6 | 977 | ||
XM_008771500.1 | XP_008769722.1 | 6 | 2734 | |||
XM_008771501.1 | XP_008769723.1 | 7 | 1177 | |||
GenBank mRNA | BC099103.1 | - | 6 | 977 | 7460148 |
