siRNA Details
| Gene Symbol: Sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012213.1 | NP_001012213.1 | 8 | 804 | ||
| XM_006253645.2 | XP_006253707.1 | 8 | 1015 | |||
| XM_008771502.1 | XP_008769724.1 | 8 | 776 | |||
| GenBank mRNA | BC078935.1 | - | 8 | 804 | 7108166 | |

Sfxn1
Genomic Map