siRNA Details
| Gene Symbol: Rnf144b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108881.1 | NP_001102351.1 | 6 | 765 | ||
| XM_006253767.2 | XP_006253829.1 | 6 | 1198 | |||
| XM_008771549.1 | XP_008769771.1 | 6 | 1203 | |||
| XM_008771550.1 | XP_008769772.1 | 6 | 1291 | |||

Rnf144b
Genomic Map