siRNA Details
| Gene Symbol: Nme8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198792.2 | NP_942087.2 | 12 | 1191 | ||
| XM_006254088.2 | XP_006254150.1 | 12 | 1240 | |||
| GenBank mRNA | AF548544.1 | - | 10 | 898 | ||
| BC097265.1 | - | 13 | 1225 | |||

Nme8
Genomic Map