siRNA Details
| Gene Symbol: Gabpb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135015.1 | NP_001128487.1 | 1 | 299 | ||
| XM_006254075.2 | XP_006254137.1 | 2 | 332 | |||
| Gene Symbol: Gabpb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039036.1 | NP_001034125.1 | 4 | 448 | ||
| XM_008762189.1 | XP_008760411.1 | 4 | 991 | |||
| XM_008762190.1 | XP_008760412.1 | 3 | 2994 | |||
| GenBank mRNA | BC098859.1 | - | 4 | 448 | 7380252 | |

Gabpb1l
Genomic Map