siRNA Details
Gene Symbol: Gabpb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135015.1 | NP_001128487.1 | 1 | 439 | ||
XM_006254075.2 | XP_006254137.1 | 2 | 472 |
Gene Symbol: Gabpb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039036.1 | NP_001034125.1 | 4 | 588 | ||
XM_008762189.1 | XP_008760411.1 | 4 | 1131 | |||
XM_008762190.1 | XP_008760412.1 | 3 | 3134 | |||
GenBank mRNA | BC098859.1 | - | 4 | 588 | 7380252 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001271492, NM_207669, NM_001271468, NM_010249, NM_001271467, and others.
Gene Ontology Categories:
