siRNA Details
| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108883.1 | NP_001102353.1 | 2 | 333 | ||
| XM_008771877.1 | XP_008770099.1 | 2 | 463 | |||
| XM_008771878.1 | XP_008770100.1 | 2 | 1537 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_022724, AK052042, AF149205, BC032960, AK083457, and others.

Suv39h2
Genomic Map