siRNA Details
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108883.1 | NP_001102353.1 | 3 | 736 | ||
XM_008771877.1 | XP_008770099.1 | 3 | 866 | |||
XM_008771878.1 | XP_008770100.1 | 3 | 1940 |
