siRNA Details
| Gene Symbol: Iba57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108827.1 | NP_001102297.1 | 3 | 1820 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Iba57| Gene Symbol: Iba57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108827.1 | NP_001102297.1 | 3 | 1820 | ||