siRNA Details
| Gene Symbol: Klhl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108885.1 | NP_001102355.1 | 5 | 1258 | ||
| XM_008771952.1 | XP_008770174.1 | 2 | 1177 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl14| Gene Symbol: Klhl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108885.1 | NP_001102355.1 | 5 | 1258 | ||
| XM_008771952.1 | XP_008770174.1 | 2 | 1177 | |||