siRNA Details
| Gene Symbol: Isoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014242.1 | NP_001014264.1 | 3 | 508 | ||
| GenBank mRNA | AB108669.1 | - | 3 | 508 | ||
| Gene Symbol: LOC103694869 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773818.1 | XP_008772040.1 | 3 | 527 | ||

Isoc1
Genomic Map