siRNA Details
| Gene Symbol: Asphd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009716.1 | NP_001009716.1 | 2 | 1312 | ||
| XM_008769345.1 | XP_008767567.1 | 3 | 2545 | |||
| XM_008769346.1 | XP_008767568.1 | 4 | 2775 | |||
| XM_008769347.1 | XP_008767569.1 | 4 | 1735 | |||
| XM_008769348.1 | XP_008767570.1 | 4 | 2448 | |||
| XM_008769349.1 | XP_008767571.1 | 4 | 2331 | |||
| XM_008769350.1 | XP_008767572.1 | 3 | 2233 | |||
| XM_008769351.1 | XP_008767573.1 | 2 | 1191 | |||
| GenBank mRNA | BC088863.1 | - | 2 | 1312 | 7313227 | |

Asphd2
Genomic Map