siRNA Details
| Gene Symbol: Best2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108895.1 | NP_001102365.1 | 4 | 543 | ||
| XM_006255283.2 | XP_006255345.1 | 3 | 677 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Best2| Gene Symbol: Best2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108895.1 | NP_001102365.1 | 4 | 543 | ||
| XM_006255283.2 | XP_006255345.1 | 3 | 677 | |||