siRNA Details
| Gene Symbol: Rnf150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191093.1 | NP_001178022.1 | 6 | 1085 | ||
| XM_008772417.1 | XP_008770639.1 | 5 | 1087 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf150| Gene Symbol: Rnf150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191093.1 | NP_001178022.1 | 6 | 1085 | ||
| XM_008772417.1 | XP_008770639.1 | 5 | 1087 | |||