siRNA Details
| Gene Symbol: Zcchc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255732.2 | XP_006255794.1 | 9 | 1444 | ||
| XM_344780.7 | XP_344781.5 | 9 | 1444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc14| Gene Symbol: Zcchc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255732.2 | XP_006255794.1 | 9 | 1444 | ||
| XM_344780.7 | XP_344781.5 | 9 | 1444 | |||