siRNA Details
| Gene Symbol: Sash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748645.3 | XP_003748693.1 | 9 | 1850 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sash1| Gene Symbol: Sash1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748645.3 | XP_003748693.1 | 9 | 1850 | ||