siRNA Details
| Gene Symbol: Samd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108901.1 | NP_001102371.1 | 2 | 768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd5| Gene Symbol: Samd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108901.1 | NP_001102371.1 | 2 | 768 | ||