siRNA Details
| Gene Symbol: Map3k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277694.1 | NP_001264623.1 | 5 | 958 | ||
| XM_008758622.1 | XP_008756844.1 | 5 | 1522 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k5| Gene Symbol: Map3k5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277694.1 | NP_001264623.1 | 5 | 958 | ||
| XM_008758622.1 | XP_008756844.1 | 5 | 1522 | |||