siRNA Details
| Gene Symbol: Zfp787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108904.1 | NP_001102374.1 | 2 | 1030 | ||
| XM_006228193.2 | XP_006228255.1 | 3 | 1194 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp787| Gene Symbol: Zfp787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108904.1 | NP_001102374.1 | 2 | 1030 | ||
| XM_006228193.2 | XP_006228255.1 | 3 | 1194 | |||