siRNA Details
| Gene Symbol: Vom1r54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008963.1 | NP_001008963.1 | 695 | |||
| GenBank mRNA | AY510374.1 | - | 695 | |||
| Gene Symbol: Vom1r53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008934.1 | NP_001008934.1 | 1 | 692 | ||
| GenBank mRNA | AY510369.1 | - | 1 | 692 | ||

Vom1r54
Genomic Map