siRNA Details
| Gene Symbol: Nanos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108908.1 | NP_001102378.1 | 1 | 258 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nanos2| Gene Symbol: Nanos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108908.1 | NP_001102378.1 | 1 | 258 | ||