siRNA Details
| Gene Symbol: LOC102548230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228505.2 | XP_006228567.1 | 3 | 1970 | ||
| XM_006228506.2 | XP_006228568.1 | 4 | 1226 | |||
| XM_006228507.2 | XP_006228569.1 | 4 | 1105 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548230| Gene Symbol: LOC102548230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228505.2 | XP_006228567.1 | 3 | 1970 | ||
| XM_006228506.2 | XP_006228568.1 | 4 | 1226 | |||
| XM_006228507.2 | XP_006228569.1 | 4 | 1105 | |||