siRNA Details
| Gene Symbol: Cnfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108909.1 | NP_001102379.1 | 3 | 371 | ||
| XM_008758960.1 | XP_008757182.1 | 2 | 417 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnfn| Gene Symbol: Cnfn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108909.1 | NP_001102379.1 | 3 | 371 | ||
| XM_008758960.1 | XP_008757182.1 | 2 | 417 | |||