siRNA Details
| Gene Symbol: Lrfn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127694.1 | NP_001121166.1 | 2 | 1754 | ||
| XM_006228626.2 | XP_006228688.1 | 5 | 3249 | |||
| XM_006228628.2 | XP_006228690.1 | 5 | 2552 | |||
| XM_008759173.1 | XP_008757395.1 | 4 | 3550 | |||
| XM_008759174.1 | XP_008757396.1 | 5 | 3641 | |||
| XM_008759175.1 | XP_008757397.1 | 5 | 2373 | |||

Lrfn1
Genomic Map