siRNA Details
| Gene Symbol: Upk1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108911.1 | NP_001102381.1 | 1, 2 | 193 | ||
| XM_006228782.2 | XP_006228844.1 | 3, 4 | 550 | |||
| XM_006228784.2 | XP_006228846.1 | 3, 4 | 550 | |||
| XM_008759176.1 | XP_008757398.1 | 2, 3 | 530 | |||

Upk1a
Genomic Map