siRNA Details
| Gene Symbol: Saa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009478.1 | NP_001009478.1 | 4 | 399 | ||
| GenBank mRNA | BC088188.1 | - | 4 | 399 | 7371403 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Saa4| Gene Symbol: Saa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009478.1 | NP_001009478.1 | 4 | 399 | ||
| GenBank mRNA | BC088188.1 | - | 4 | 399 | 7371403 | |