siRNA Details
| Gene Symbol: Hmgn5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134706.1 | NP_001128178.1 | 4 | 272 | ||
| GenBank mRNA | BC168165.1 | - | 4 | 174 | 8370295 | |
| BF289419.1 | - | 4 | 289 | |||
| Gene Symbol: Hmgn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276434.1 | NP_001263363.1 | 4 | 272 | ||
| GenBank mRNA | DY318327.1 | - | 4 | 171 | ||

Hmgn5b
Genomic Map