siRNA Details
| Gene Symbol: Asb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108915.1 | NP_001102385.1 | 5 | 1152 | ||
| XM_006229349.2 | XP_006229411.1 | 6 | 1354 | |||
| XM_006229350.2 | XP_006229412.1 | 6 | 1320 | |||
| XM_008759545.1 | XP_008757767.1 | 5 | 2298 | |||

Asb7
Genomic Map