siRNA Details
| Gene Symbol: Crtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229462.2 | XP_006229524.1 | 15 | 1977 | ||
| XM_006229463.2 | XP_006229525.1 | 15 | 1974 | |||
| Gene Symbol: LOC100909455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748889.3 | XP_003748937.2 | 1640 | |||
| XM_006229461.2 | XP_006229523.1 | 1638 | ||||

Crtc3
Genomic Map