siRNA Details
Gene Symbol: Crtc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229462.2 | XP_006229524.1 | 14 | 1785 | ||
XM_006229463.2 | XP_006229525.1 | 14 | 1785 |
Gene Symbol: LOC100909455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748889.3 | XP_003748937.2 | 1448 | |||
XM_006229461.2 | XP_006229523.1 | 1449 |
