siRNA Details
| Gene Symbol: LOC103690138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759595.1 | XP_008757817.1 | 2 | 3404 | ||
| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108916.1 | NP_001102386.1 | 5 | 2824 | ||

LOC103690138
Genomic Map