siRNA Details
Gene Symbol: Vom2r43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099512.1 | NP_001092982.1 | 3 | 765 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
